siRNA Details
| Gene Symbol: OR2T12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004692.1 | NP_001004692.1 | 1 | 249 | ||
| Gene Symbol: OR2T33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004695.1 | NP_001004695.1 | 1 | 249 | ||
| GenBank mRNA | BC136994.1 | - | 1 | 271 | 9020986 | |
| BC136995.1 | - | 1 | 271 | 9020987 | ||
| Gene Symbol: OR2T8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005522.1 | NP_001005522.1 | 1 | 249 | ||
| XM_011544178.2 | XP_011542480.1 | 2 | 1592 | |||
| XM_017001172.1 | XP_016856661.1 | 2 | 4371 | |||

OR2T12
Genomic Map