siRNA Details
| Gene Symbol: TYW3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162916.1 | NP_001156388.1 | 5 | 689 | ||
| NM_138467.2 | NP_612476.1 | 6 | 784 | |||
| NR_027962.1 | 6 | 875 | ||||
| XM_006710347.1 | XP_006710410.1 | 6 | 797 | |||
| GenBank mRNA | AK098237.1 | - | 6 | 784 | ||
| AK294264.1 | - | 3 | 514 | |||
| AK299849.1 | - | 6 | 640 | |||
| AK311272.1 | - | 6 | 821 | |||
| BC032341.2 | - | 1 | 136 | |||
| BC071765.1 | - | 6 | 784 | 3931600 | ||
| BX647591.1 | - | 2 | 231 | |||

TYW3
Genomic Map