siRNA Details
| Gene Symbol: TPRG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182752.3 | NP_877429.2 | 5 | 851 | ||
| GenBank mRNA | AK025464.1 | - | 4 | 571 | ||
| AK096427.1 | - | 5 | 828 | |||
| AK289919.1 | - | 5 | 801 | |||
| AK298343.1 | - | 5 | 690 | |||
| BC019034.2 | - | 4 | 616 | 4576259 | ||

TPRG1L
Genomic Map