siRNA Details
| Gene Symbol: PODN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153703.4 | NP_714914.2 | 5 | 851 | ||
| XM_005270457.3 | XP_005270514.1 | 6 | 880 | |||
| XM_017000294.1 | XP_016855783.1 | 5 | 1201 | |||
| GenBank mRNA | AK296649.1 | - | 6 | 746 | ||
| AK299375.1 | - | 7 | 936 | |||
| AK314096.1 | - | 5 | 445 | |||
| AY313607.1 | - | 5 | 690 | |||
| AY358294.1 | - | 7 | 967 | |||
| BC028292.1 | - | 5 | 674 | 4938380 | ||
| BC030608.2 | - | 5 | 692 | 4826932 | ||

PODN
Genomic Map