siRNA Details
| Gene Symbol: DCST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144622.2 | NP_653223.2 | 8 | 1314 | ||
| XM_011509188.2 | XP_011507490.1 | 6 | 777 | |||
| XM_011509189.2 | XP_011507491.1 | 3 | 1166 | |||
| GenBank mRNA | AK057496.1 | - | 8 | 1314 | ||
| AK097468.1 | - | 6 | 728 | |||
| AK097993.1 | - | 3 | 986 | |||
| BC111794.1 | - | 8 | 1293 | |||

DCST2
Genomic Map