siRNA Details
| Gene Symbol: KLF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173484.3 | NP_775755.3 | 2 | 386 | ||
| XM_011540700.1 | XP_011539002.1 | 5 | 1082 | |||
| GenBank mRNA | AK097479.1 | - | 2 | 358 | ||
| AK308458.1 | - | 4 | 916 | |||
| BC035408.1 | - | 2 | 343 | |||
| BC049844.1 | - | 2 | 386 | 5166200 | ||
| DB046781.1 | - | 2 | 358 | |||

KLF17
Genomic Map