siRNA Details
| Gene Symbol: TMEM125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320244.1 | NP_001307173.1 | 4 | 1389 | ||
| NM_144626.2 | NP_653227.1 | 4 | 1294 | |||
| XM_005270467.3 | XP_005270524.1 | 3 | 1534 | |||
| XM_011540705.2 | XP_011539007.1 | 4 | 1520 | |||
| GenBank mRNA | BC016858.1 | - | 3 | 1147 | 3845811 | |
| BC072393.1 | - | 4 | 1377 | |||

TMEM125
Genomic Map