siRNA Details
| Gene Symbol: C1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300834.1 | NP_001287763.1 | 6 | 917 | ||
| NM_001300835.1 | NP_001287764.1 | 5 | 897 | |||
| NM_174896.3 | NP_777556.1 | 6 | 992 | |||
| XM_011540712.2 | XP_011539014.1 | 7 | 6472 | |||
| XM_017000323.1 | XP_016855812.1 | 7 | 2266 | |||
| XM_017000324.1 | XP_016855813.1 | 6 | 6377 | |||
| GenBank mRNA | AK123160.1 | - | 3 | 2271 | ||
| BC017973.1 | - | 6 | 879 | 4693393 | ||

C1orf162
Genomic Map