siRNA Details
| Gene Symbol: C1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300834.1 | NP_001287763.1 | 3 | 260 | ||
| NM_001300835.1 | NP_001287764.1 | 3 | 260 | |||
| NM_174896.3 | NP_777556.1 | 3 | 260 | |||
| XM_011540712.2 | XP_011539014.1 | 4 | 5740 | |||
| XM_017000323.1 | XP_016855812.1 | 4 | 1609 | |||
| XM_017000324.1 | XP_016855813.1 | 4 | 5740 | |||
| GenBank mRNA | AK123160.1 | - | 2 | 1548 | ||
| BC017973.1 | - | 3 | 147 | 4693393 | ||
| BG545926.1 | - | 3 | 150 | |||
| HX785231.1 | - | 3 | 142 | |||

C1orf162
Genomic Map