siRNA Details
Gene Symbol: SPATA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080608.3 | NP_542175.1 | 2 | 258 | ||
GenBank mRNA | BC039607.1 | - | 2 | 258 | 5167905 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPATA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080608.3 | NP_542175.1 | 2 | 258 | ||
GenBank mRNA | BC039607.1 | - | 2 | 258 | 5167905 |