siRNA Details
| Gene Symbol: TSC22D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015881.1 | NP_001015881.1 | 3 | 1115 | ||
| NM_001318468.1 | NP_001305397.1 | 4 | 1583 | |||
| NM_001318470.1 | NP_001305399.1 | 4 | 1569 | |||
| NM_004089.3 | NP_004080.2 | 3 | 1320 | |||
| NM_198057.2 | NP_932174.1 | 3 | 1636 | |||
| XM_005262099.1 | XP_005262156.1 | 5 | 1597 | |||
| XM_005262100.1 | XP_005262157.1 | 4 | 1472 | |||
| XM_005262102.1 | XP_005262159.1 | 4 | 1465 | |||
| XM_005262103.3 | XP_005262160.1 | 4 | 1575 | |||
| XM_011530884.1 | XP_011529186.1 | 4 | 1671 | |||
| XM_017029335.1 | XP_016884824.1 | 4 | 1407 | |||
| GenBank mRNA | AB025432.1 | - | 3 | 1304 | ||
| AF183393.1 | - | 3 | 1306 | |||
| AF228339.1 | - | 3 | 1310 | |||
| AK092645.1 | - | 4 | 1522 | |||
| AK092669.1 | - | 3 | 1608 | |||
| AK127938.1 | - | 3 | 1260 | |||
| AL110191.1 | - | 3 | 1318 | |||
| AY007119.1 | - | 3 | 1219 | |||
| BC018148.1 | - | 3 | 1002 | |||
| BC072446.1 | - | 3 | 1314 | 6165765 | ||
| BX647854.1 | - | 4 | 1569 | |||
| CR933650.1 | - | 3 | 1322 | |||

TSC22D3
Genomic Map