siRNA Details
| Gene Symbol: CSTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138283.1 | NP_612140.1 | 2 | 398 | ||
| GenBank mRNA | AY707749.1 | - | 2 | 398 | ||
| BC117396.1 | - | 1 | 198 | 40125947 | ||
| BI560738.1 | - | 2 | 404 | |||
| FJ895879.1 | - | 1 | 23 | |||
| FJ895880.1 | - | 1 | 23 | |||

CSTL1
Genomic Map