siRNA Details
| Gene Symbol: CST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008693.2 | NP_001008693.2 | 1 | 236 | ||
| GenBank mRNA | AF494536.1 | - | 1 | 139 | ||
| BC137302.1 | - | 1 | 236 | 9021299 | ||
| BC137303.1 | - | 1 | 236 | 9021300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST9| Gene Symbol: CST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008693.2 | NP_001008693.2 | 1 | 236 | ||
| GenBank mRNA | AF494536.1 | - | 1 | 139 | ||
| BC137302.1 | - | 1 | 236 | 9021299 | ||
| BC137303.1 | - | 1 | 236 | 9021300 | ||