siRNA Details
| Gene Symbol: C20orf144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080825.3 | NP_543015.1 | 1 | 150 | ||
| GenBank mRNA | BC032818.1 | - | 1 | 100 | 5172528 | |
| BX378327.2 | - | 1 | 150 | |||
| DQ080434.1 | - | 1 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf144| Gene Symbol: C20orf144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080825.3 | NP_543015.1 | 1 | 150 | ||
| GenBank mRNA | BC032818.1 | - | 1 | 100 | 5172528 | |
| BX378327.2 | - | 1 | 150 | |||
| DQ080434.1 | - | 1 | 152 | |||