siRNA Details
| Gene Symbol: FAM83C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178468.5 | NP_848563.1 | 4 | 1408 | ||
| GenBank mRNA | AK091116.1 | - | 5 | 975 | ||
| AK091138.1 | - | 3 | 751 | |||
| BC113483.1 | - | 4 | 1387 | 8322535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM83C| Gene Symbol: FAM83C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178468.5 | NP_848563.1 | 4 | 1408 | ||
| GenBank mRNA | AK091116.1 | - | 5 | 975 | ||
| AK091138.1 | - | 3 | 751 | |||
| BC113483.1 | - | 4 | 1387 | 8322535 | ||