siRNA Details
| Gene Symbol: C22orf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173793.4 | NP_776154.3 | 3 | 1599 | ||
| GenBank mRNA | AK095271.1 | - | 4 | 595 | ||
| BC062599.1 | - | 3 | 1064 | 5248209 | ||
| BX648073.1 | - | 1 | 368 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22orf39| Gene Symbol: C22orf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173793.4 | NP_776154.3 | 3 | 1599 | ||
| GenBank mRNA | AK095271.1 | - | 4 | 595 | ||
| BC062599.1 | - | 3 | 1064 | 5248209 | ||
| BX648073.1 | - | 1 | 368 | |||