siRNA Details
| Gene Symbol: C22orf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173793.4 | NP_776154.3 | 3 | 1188 | ||
| GenBank mRNA | BC062599.1 | - | 3 | 653 | 5248209 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22orf39| Gene Symbol: C22orf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173793.4 | NP_776154.3 | 3 | 1188 | ||
| GenBank mRNA | BC062599.1 | - | 3 | 653 | 5248209 | |