siRNA Details
| Gene Symbol: XIRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079810.3 | NP_001073278.1 | 10 | 2412 | ||
| NM_001199143.1 | NP_001186072.1 | 11 | 2511 | |||
| NM_001199144.1 | NP_001186073.1 | 9 | 11389 | |||
| NM_001199145.1 | NP_001186074.1 | 8 | 2010 | |||
| NM_152381.5 | NP_689594.4 | 11 | 11791 | |||
| XM_017003309.1 | XP_016858798.1 | 9 | 11286 | |||
| GenBank mRNA | AK122889.1 | - | 9 | 1851 | ||
| AK123963.1 | - | 1 | 199 | |||
| AL831970.2 | - | 3 | 3993 | |||
| AL831985.1 | - | 3 | 3653 | |||
| AL832011.1 | - | 3 | 4277 | |||
| AL832336.1 | - | 3 | 4318 | |||
| AL832360.1 | - | 3 | 3898 | |||
| AL832375.1 | - | 3 | 3328 | |||
| AL832382.1 | - | 3 | 3845 | |||
| AL832452.2 | - | 4 | 4366 | |||
| AL833291.2 | - | 4 | 5609 | |||
| BX647448.1 | - | 3 | 5341 | |||
| BX647645.1 | - | 3 | 4012 | |||
| BX647845.1 | - | 3 | 3950 | |||
| EF119711.1 | - | 9 | 11389 | |||
| EF119717.1 | - | 8 | 2010 | |||
| EF119718.1 | - | 12 | 2611 | |||
| EF119719.1 | - | 10 | 2513 | |||
| EF119720.1 | - | 10 | 2412 | |||
| EF119721.1 | - | 11 | 2612 | |||
| EF119722.1 | - | 11 | 2511 | |||

XIRP2
Genomic Map