siRNA Details
| Gene Symbol: TMEM18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152834.2 | NP_690047.2 | 3, 4 | 322 | ||
| GenBank mRNA | AK125009.1 | - | 2, 3 | 1207 | ||
| AL137269.1 | - | 3, 4 | 322 | |||
| BC016010.1 | - | 3, 4 | 294 | 4686355 | ||
| BC032379.1 | - | 3, 4 | 582 | 5109444 | ||

TMEM18
Genomic Map