siRNA Details
Gene Symbol: TMEM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152834.2 | NP_690047.2 | 3, 4 | 332 | ||
GenBank mRNA | AK125009.1 | - | 2, 3 | 1217 | ||
AL137269.1 | - | 3, 4 | 332 | |||
BC016010.1 | - | 3, 4 | 304 | 4686355 | ||
BC032379.1 | - | 3, 4 | 592 | 5109444 |
