siRNA Details
| Gene Symbol: REG3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008387.2 | NP_001008388.1 | 4 | 485 | ||
| NM_198448.3 | NP_940850.1 | 4 | 385 | |||
| XM_005264135.2 | XP_005264192.1 | 4 | 547 | |||
| GenBank mRNA | AB161037.1 | - | 4 | 370 | ||
| AB161038.1 | - | 4 | 470 | |||
| AK292595.1 | - | 4 | 369 | |||
| AY359047.1 | - | 4 | 371 | |||
| AY428734.1 | - | 4 | 344 | |||
| BC103853.1 | - | 4 | 444 | 40002929 | ||
| BC103854.1 | - | 4 | 344 | 40002931 | ||
| BX280299.1 | - | 4 | 385 | |||
| CR541938.1 | - | 3 | 286 | |||

REG3G
Genomic Map