siRNA Details
| Gene Symbol: PLEKHH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172069.3 | NP_742066.2 | 29 | 4315 | ||
| XM_017003351.1 | XP_016858840.1 | 28 | 4302 | |||
| XM_017003353.1 | XP_016858842.1 | 30 | 4426 | |||
| GenBank mRNA | AB095948.1 | - | 27 | 4101 | ||
| AL832207.1 | - | 29 | 4308 | |||
| AL833400.1 | - | 22 | 2618 | |||
| AL833473.1 | - | 4 | 450 | |||
| AL833546.1 | - | 29 | 6244 | |||

PLEKHH2
Genomic Map