siRNA Details
| Gene Symbol: KCTD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321547.1 | NP_001308476.1 | 7 | 956 | ||
| NM_001321548.1 | NP_001308477.1 | 7 | 934 | |||
| NM_001321550.1 | NP_001308479.1 | 7 | 1317 | |||
| NM_152387.3 | NP_689600.2 | 7 | 1339 | |||
| GenBank mRNA | AK095137.1 | - | 1 | 368 | ||
| AK290867.1 | - | 7 | 930 | |||
| BC045189.1 | - | 7 | 1310 | 5296647 | ||
| BC059366.1 | - | 7 | 1287 | |||
| BC067755.1 | - | 7 | 930 | 5299303 | ||
| BU181789.1 | - | 3 | 195 | |||

KCTD18
Genomic Map