siRNA Details
| Gene Symbol: KCTD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321547.1 | NP_001308476.1 | 6 | 882 | ||
| NM_001321548.1 | NP_001308477.1 | 6 | 860 | |||
| NM_001321550.1 | NP_001308479.1 | 6 | 1243 | |||
| NM_152387.3 | NP_689600.2 | 6 | 1265 | |||
| GenBank mRNA | AK290867.1 | - | 6 | 856 | ||
| BC045189.1 | - | 6 | 1236 | 5296647 | ||
| BC059366.1 | - | 6 | 1213 | |||
| BC067755.1 | - | 6 | 856 | 5299303 | ||
| BU181789.1 | - | 2 | 121 | |||

KCTD18
Genomic Map