siRNA Details
| Gene Symbol: KCTD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321547.1 | NP_001308476.1 | 4 | 621 | ||
| NM_001321548.1 | NP_001308477.1 | 4 | 599 | |||
| NM_001321550.1 | NP_001308479.1 | 4 | 982 | |||
| NM_152387.3 | NP_689600.2 | 4 | 1004 | |||
| GenBank mRNA | AK055884.1 | - | 4 | 596 | ||
| AK290867.1 | - | 4 | 595 | |||
| BC045189.1 | - | 4 | 975 | 5296647 | ||
| BC059366.1 | - | 4 | 952 | |||
| BC067755.1 | - | 4 | 595 | 5299303 | ||
| CB960352.1 | - | 4 | 622 | |||

KCTD18
Genomic Map