siRNA Details
| Gene Symbol: TMEM182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321343.1 | NP_001308272.1 | 5 | 509 | ||
| NM_001321344.1 | NP_001308273.1 | 4 | 382 | |||
| NM_001321345.1 | NP_001308274.1 | 3 | 297 | |||
| NM_001321346.1 | NP_001308275.1 | 3 | 297 | |||
| NM_144632.4 | NP_653233.4 | 3 | 508 | |||
| XM_006712287.1 | XP_006712350.1 | 3 | 506 | |||
| XM_006712288.3 | XP_006712351.1 | 3 | 545 | |||
| XM_011510632.2 | XP_011508934.1 | 3 | 506 | |||
| XM_017003375.1 | XP_016858864.1 | 3 | 545 | |||
| XM_017003376.1 | XP_016858865.1 | 3 | 293 | |||
| GenBank mRNA | AK054856.1 | - | 3 | 268 | ||
| AK129862.1 | - | 3 | 467 | |||
| AL832688.1 | - | 4 | 379 | |||
| AY358262.1 | - | 3 | 606 | |||
| BC073844.1 | - | 3 | 244 | |||
| BC107694.1 | - | 3 | 252 | |||
| BC130306.1 | - | 3 | 455 | 40146337 | ||
| BC130308.1 | - | 3 | 452 | 40146339 | ||
| CR936731.1 | - | 3 | 484 | |||
| CR936732.1 | - | 3 | 480 | |||
| CR936735.1 | - | 3 | 507 | |||
| CX163622.1 | - | 3 | 237 | |||
| DA355461.1 | - | 3 | 295 | |||
| DA908857.1 | - | 5 | 507 | |||
| HY012093.1 | - | 3 | 297 | |||

TMEM182
Genomic Map