siRNA Details
| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017928.3 | NP_001017928.1 | 4 | 357 | ||
| NM_001308326.1 | NP_001295255.1 | 4 | 383 | |||
| XM_011512410.2 | XP_011510712.1 | 4 | 335 | |||
| GenBank mRNA | BC054051.1 | - | 4 | 348 | ||
| BC062725.1 | - | 4 | 357 | 6495960 | ||
| BC109378.1 | - | 4 | 341 | |||
| CD683826.1 | - | 4 | 379 | |||
| DA025111.1 | - | 4 | 379 | |||

CCDC58
Genomic Map