siRNA Details
| Gene Symbol: LYZL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304386.1 | NP_001291315.1 | 4 | 602 | ||
| NM_144634.3 | NP_653235.1 | 4 | 621 | |||
| XM_011533355.2 | XP_011531657.1 | 4 | 664 | |||
| XM_017005705.1 | XP_016861194.1 | 6 | 976 | |||
| XM_017005706.1 | XP_016861195.1 | 5 | 734 | |||
| GenBank mRNA | AF326749.1 | - | 4 | 550 | ||
| BC016747.2 | - | 4 | 592 | 4830396 | ||

LYZL4
Genomic Map