siRNA Details
| Gene Symbol: KLHL40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152393.3 | NP_689606.2 | 6 | 1956 | ||
| GenBank mRNA | AK300682.1 | - | 7 | 1262 | ||
| AY176040.1 | - | 6 | 1956 | |||
| AY177390.1 | - | 6 | 1956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL40| Gene Symbol: KLHL40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152393.3 | NP_689606.2 | 6 | 1956 | ||
| GenBank mRNA | AK300682.1 | - | 7 | 1262 | ||
| AY176040.1 | - | 6 | 1956 | |||
| AY177390.1 | - | 6 | 1956 | |||