siRNA Details
| Gene Symbol: FAM131A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024378.1 | - | 1 | 2510 | ||
| AY358435.1 | - | 1 | 1497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM131A| Gene Symbol: FAM131A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024378.1 | - | 1 | 2510 | ||
| AY358435.1 | - | 1 | 1497 | |||