siRNA Details
| Gene Symbol: DCBLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080927.3 | NP_563615.3 | 16 | 2324 | ||
| XM_011512419.2 | XP_011510721.1 | 15 | 2040 | |||
| GenBank mRNA | AB073146.1 | - | 16 | 2324 | ||
| AF387547.1 | - | 16 | 1972 | |||
| AK302649.1 | - | 16 | 1804 | |||
| BC029658.1 | - | 16 | 1885 | 4431275 | ||

DCBLD2
Genomic Map