siRNA Details
| Gene Symbol: LRRC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135057.2 | NP_001128529.2 | 3 | 1695 | ||
| NM_130830.4 | NP_570843.2 | 2 | 1638 | |||
| GenBank mRNA | AB071037.1 | - | 1 | 1552 | ||
| AK125112.1 | - | 2 | 1638 | |||
| BC101064.1 | - | 3 | 1683 | 40008605 | ||
| BC101065.1 | - | 2 | 1626 | 40008610 | ||

LRRC15
Genomic Map