siRNA Details
| Gene Symbol: FAM43A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153690.4 | NP_710157.2 | 1 | 1133 | ||
| GenBank mRNA | AF289585.1 | - | 1 | 787 | ||
| AK074503.1 | - | 1 | 299 | |||
| BC039689.2 | - | 1 | 342 | 4906944 | ||
| BC064989.1 | - | 1 | 734 | 5423287 | ||

FAM43A
Genomic Map