siRNA Details
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207316.2 | NP_997199.1 | 1 | 110 | ||
GenBank mRNA | AY358607.1 | - | 1 | 110 | ||
BC071780.1 | - | 1 | 101 | 4607584 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207316.2 | NP_997199.1 | 1 | 110 | ||
GenBank mRNA | AY358607.1 | - | 1 | 110 | ||
BC071780.1 | - | 1 | 101 | 4607584 |