siRNA Details
| Gene Symbol: C3orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026866.1 | 3 | 646 | |||
| GenBank mRNA | BC015210.1 | - | 3 | 615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C3orf49| Gene Symbol: C3orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026866.1 | 3 | 646 | |||
| GenBank mRNA | BC015210.1 | - | 3 | 615 | ||