siRNA Details
| Gene Symbol: OCIAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014446.2 | NP_001014446.1 | 3 | 327 | ||
| NM_001286773.1 | NP_001273702.1 | 3 | 350 | |||
| NM_152398.3 | NP_689611.1 | 3 | 327 | |||
| GenBank mRNA | AK298305.1 | - | 3 | 296 | ||
| AY472117.1 | - | 3 | 329 | |||
| BC032808.1 | - | 3 | 298 | 4454708 | ||
| BG748993.1 | - | 3 | 234 | |||
| BI546814.1 | - | 3 | 332 | |||
| BM782144.1 | - | 3 | 297 | |||
| CR749310.1 | - | 3 | 272 | |||
| W68799.1 | - | 3 | 279 | |||

OCIAD2
Genomic Map