siRNA Details
| Gene Symbol: SCLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144643.3 | NP_653244.2 | 11 | 1347 | ||
| XM_017007716.1 | XP_016863205.1 | 11 | 1347 | |||
| XM_017007717.1 | XP_016863206.1 | 11 | 1347 | |||
| XM_017007718.1 | XP_016863207.1 | 11 | 1347 | |||
| XM_017007719.1 | XP_016863208.1 | 11 | 868 | |||
| XM_017007720.1 | XP_016863209.1 | 12 | 1259 | |||
| GenBank mRNA | AK055217.1 | - | 11 | 1347 | ||
| AK093361.1 | - | 4 | 310 | |||

SCLT1
Genomic Map