siRNA Details
| Gene Symbol: TMEM155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317837.1 | NP_001304766.1 | 4, 5 | 467 | ||
| NM_001317838.1 | NP_001304767.1 | 4, 5 | 464 | |||
| NM_001317839.1 | NP_001304768.1 | 4, 5 | 342 | |||
| NM_152399.3 | NP_689612.3 | 4, 5 | 623 | |||
| XM_005262737.2 | XP_005262794.1 | 4, 5 | 431 | |||
| XM_017007725.1 | XP_016863214.1 | 4, 5 | 337 | |||
| GenBank mRNA | AK055396.1 | - | 4, 5 | 344 | ||
| AL552582.3 | - | 4, 5 | 494 | |||
| BC047657.1 | - | 4, 5 | 623 | 5264721 | ||
| DA176165.1 | - | 4, 5 | 428 | |||

TMEM155
Genomic Map