siRNA Details
| Gene Symbol: ZFP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304358.1 | NP_001291287.1 | 3 | 674 | ||
| NM_174900.4 | NP_777560.2 | 4 | 840 | |||
| XM_011531604.2 | XP_011529906.1 | 3 | 695 | |||
| XM_011531605.1 | XP_011529907.1 | 4 | 845 | |||
| XM_011531606.1 | XP_011529908.1 | 4 | 849 | |||
| XM_011531607.2 | XP_011529909.1 | 3 | 692 | |||
| GenBank mRNA | AF450454.1 | - | 1 | 432 | ||
| AK056719.1 | - | 3 | 674 | |||
| AY429593.1 | - | 4 | 840 | |||

ZFP42
Genomic Map