siRNA Details
| Gene Symbol: SPATA18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145263.3 | NP_660306.1 | 4, 5 | 793 | ||
| NR_123728.1 | 4, 5 | 793 | ||||
| XM_005265717.2 | XP_005265774.1 | 4, 5 | 708 | |||
| XM_017007731.1 | XP_016863220.1 | 2, 3 | 191 | |||
| GenBank mRNA | AB465501.1 | - | 4, 5 | 419 | ||
| AK057468.1 | - | 4, 5 | 705 | |||
| AK098625.1 | - | 1, 2 | 18 | |||
| AK314799.1 | - | 4, 5 | 663 | |||
| BC025396.1 | - | 4, 5 | 700 | 4821894 | ||

SPATA18
Genomic Map