siRNA Details
| Gene Symbol: SPATA18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297608.1 | NP_001284537.1 | 2 | 501 | ||
| NM_145263.3 | NP_660306.1 | 2 | 501 | |||
| NR_123728.1 | 2 | 501 | ||||
| XM_005265716.1 | XP_005265773.1 | 2 | 416 | |||
| XM_005265717.2 | XP_005265774.1 | 2 | 416 | |||
| XM_011534369.1 | XP_011532671.1 | 2 | 416 | |||
| XM_011534370.1 | XP_011532672.1 | 2 | 416 | |||
| XM_017007731.1 | XP_016863220.1 | 1 | 15 | |||
| GenBank mRNA | AB465501.1 | - | 2 | 127 | ||
| AB465502.1 | - | 2 | 127 | |||
| AK057468.1 | - | 2 | 413 | |||
| AK304386.1 | - | 2 | 416 | |||
| AK314799.1 | - | 2 | 371 | |||
| BC025396.1 | - | 2 | 408 | 4821894 | ||
| DC404003.1 | - | 2 | 501 | |||

SPATA18
Genomic Map