siRNA Details
| Gene Symbol: SPATA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144644.3 | NP_653245.2 | 3 | 558 | ||
| GenBank mRNA | AY040204.3 | - | 3 | 558 | ||
| BC021731.2 | - | 3 | 459 | 5269042 | ||
| BC039342.1 | - | 2 | 1526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA4| Gene Symbol: SPATA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144644.3 | NP_653245.2 | 3 | 558 | ||
| GenBank mRNA | AY040204.3 | - | 3 | 558 | ||
| BC021731.2 | - | 3 | 459 | 5269042 | ||
| BC039342.1 | - | 2 | 1526 | |||