siRNA Details
| Gene Symbol: AASDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286668.1 | NP_001273597.1 | 5 | 981 | ||
| NM_001286669.1 | NP_001273598.1 | 5 | 1132 | |||
| NM_001286670.1 | NP_001273599.1 | 5 | 936 | |||
| NM_001286671.1 | NP_001273600.1 | 6 | 1253 | |||
| NM_001286672.1 | NP_001273601.1 | 6 | 1253 | |||
| NM_001323890.1 | NP_001310819.1 | 6 | 1253 | |||
| NM_001323892.1 | NP_001310821.1 | 6 | 1253 | |||
| NM_001323893.1 | NP_001310822.1 | 5 | 1132 | |||
| NM_001323899.1 | NP_001310828.1 | 6 | 1253 | |||
| NM_181806.3 | NP_861522.2 | 6 | 1253 | |||
| XM_017007740.1 | XP_016863229.1 | 6 | 1214 | |||
| XM_017007741.1 | XP_016863230.1 | 7 | 1329 | |||
| XM_017007742.1 | XP_016863231.1 | 4 | 1029 | |||
| XM_017007743.1 | XP_016863232.1 | 6 | 1214 | |||
| XM_017007744.1 | XP_016863233.1 | 3 | 836 | |||
| GenBank mRNA | AF516672.1 | - | 5 | 896 | ||
| AK294332.1 | - | 5 | 898 | |||
| AK303733.1 | - | 5 | 1085 | |||
| AK304309.1 | - | 6 | 1244 | |||
| AK316296.1 | - | 6 | 1234 | |||
| AY314787.1 | - | 6 | 1216 | |||
| AY422212.1 | - | 6 | 1168 | |||
| BC141824.1 | - | 6 | 1211 | 40148174 | ||
| BC142709.1 | - | 6 | 1189 | 40148820 | ||
| BX640635.1 | - | 4 | 780 | |||
| BX648853.1 | - | 6 | 1245 | |||

AASDH
Genomic Map