siRNA Details
| Gene Symbol: C4orf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144645.3 | NP_653246.1 | 4 | 598 | ||
| XM_011531610.2 | XP_011529912.1 | 4 | 485 | |||
| XM_011531611.2 | XP_011529913.1 | 4 | 707 | |||
| XM_011531612.2 | XP_011529914.1 | 4 | 417 | |||
| XM_011531613.2 | XP_011529915.1 | 4 | 952 | |||
| XM_011531615.2 | XP_011529917.1 | 4 | 437 | |||
| XM_017007746.1 | XP_016863235.1 | 4 | 379 | |||
| GenBank mRNA | BC016746.1 | - | 4 | 574 | 4827131 | |
| BX090513.1 | - | 3 | 339 | |||

C4orf36
Genomic Map