siRNA Details
| Gene Symbol: OTOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177998.1 | NP_819056.1 | 5 | 1147 | ||
| GenBank mRNA | BC130430.1 | - | 5 | 1142 | 40146461 | |
| BC130432.1 | - | 5 | 1142 | 40146463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTOP1| Gene Symbol: OTOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177998.1 | NP_819056.1 | 5 | 1147 | ||
| GenBank mRNA | BC130430.1 | - | 5 | 1142 | 40146461 | |
| BC130432.1 | - | 5 | 1142 | 40146463 | ||