siRNA Details
| Gene Symbol: C5orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144954.1 | NP_001138426.1 | 1, 2 | 432 | ||
| XM_011534431.1 | XP_011532733.1 | 1, 2 | 426 | |||
| XM_017009028.1 | XP_016864517.1 | 1, 2 | 426 | |||
| GenBank mRNA | BC034971.1 | - | 1, 2 | 398 | ||
| BC047225.1 | - | 1, 2 | 387 | |||
| BC053953.1 | - | 1, 2 | 387 | |||
| BC057842.1 | - | 1, 2 | 373 | |||
| BC066964.1 | - | 1, 2 | 394 | |||
| BC092489.1 | - | 1, 2 | 421 | |||
| DB443869.1 | - | 1, 2 | 432 | |||

C5orf47
Genomic Map