siRNA Details
| Gene Symbol: MROH2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173489.4 | NP_775760.3 | 30 | 3587 | ||
| XM_011513952.1 | XP_011512254.1 | 30 | 3573 | |||
| XM_011513953.1 | XP_011512255.1 | 29 | 3511 | |||
| GenBank mRNA | AK097562.1 | - | 6 | 633 | ||
| AK126005.1 | - | 15 | 1456 | |||
| AK128183.1 | - | 20 | 2053 | |||
| AL832763.1 | - | 25 | 2512 | |||
| BC044937.1 | - | 18 | 2059 | |||
| BC052962.1 | - | 30 | 3548 | 4838071 | ||
| CR749346.1 | - | 25 | 2488 | |||

MROH2B
Genomic Map