siRNA Details
| Gene Symbol: FAM173B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK302012.1 | - | 2 | 254 | ||
| BC063406.1 | - | 2 | 233 | 5190223 | ||
| BG611454.1 | - | 2 | 245 | |||
| DA843869.1 | - | 2 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM173B| Gene Symbol: FAM173B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK302012.1 | - | 2 | 254 | ||
| BC063406.1 | - | 2 | 233 | 5190223 | ||
| BG611454.1 | - | 2 | 245 | |||
| DA843869.1 | - | 2 | 241 | |||