siRNA Details
| Gene Symbol: AFAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323062.1 | NP_001309991.1 | 17 | 2233 | ||
| NM_152406.3 | NP_689619.1 | 18 | 2332 | |||
| XM_011537558.1 | XP_011535860.1 | 14 | 1882 | |||
| XM_017009036.1 | XP_016864525.1 | 18 | 2309 | |||
| GenBank mRNA | AK074185.1 | - | 18 | 2275 | ||
| AK094067.1 | - | 9 | 1362 | |||
| BC125093.1 | - | 18 | 2251 | 40118258 | ||

AFAP1L1
Genomic Map