siRNA Details
| Gene Symbol: TMEM171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161342.2 | NP_001154814.1 | 2 | 254 | ||
| NM_173490.7 | NP_775761.4 | 2 | 254 | |||
| XM_011543156.1 | XP_011541458.1 | 2 | 234 | |||
| GenBank mRNA | BC018083.1 | - | 2 | 240 | 4797244 | |
| BC035310.1 | - | 1 | 95 | 4746772 | ||
| CN261444.1 | - | 2 | 257 | |||

TMEM171
Genomic Map